missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum Trioxide, TRC
Specifications
Specifications
| Chemical Name or Material | Molybdenum trioxide |
| CAS | 1313-27-5 |
| Quantity | 25 g |
| Molecular Formula | MoO3 |
| Synonym | Molybdenum oxide (MoO3), Bouen SKN 301, JCPDS 35-0609, MFR, MOP-P 100, Mo 1202T, Molybdena, Molybdenum oxide, Molybdenum oxide (Mo4O12), Molybdenum oxide (Mo5O15), Molybdenum trioxide, Molybdenum trioxide pentamer, Molybdenum trioxide tetramer, Molybdenum(VI) oxide, Molybdenum(VI) trioxide, Molybdic acid anhydride, Molybdic anhydride, Molybdic oxide, NSC 215191, Pol-U, SS-M 0100 |
| InChI Formula | InChI=1S/Mo.3O |
| SMILES | O=[Mo](=O)=O |
| IUPAC Name | trioxomolybdenum |
| Molecular Weight (g/mol) | 143.95 |
| Formula Weight | 145.8902 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?