missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Molybdenum Trioxide, ≥99.5%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11462882 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462882 500 g 500g
11452882 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462882 Supplier MP Biomedicals Supplier No. 0215254890
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Color Yellow
Quantity 500 g
Assay Percent Range ≥99.5%
Linear Formula MoO3
Formula Weight 143.94
Grade ACS Reagent
Concentration or Composition (by Analyte or Components) Assay (MoO3) - >99.5%, Insoluble Matter in dilute NH4OH - <0.0100%, Ammonium (NH4) - <0.0020%, Arsenate and Silicate (as SiO2) - <0.0010%, Chloride (Cl) - <0.0020%, Heavy Metals (as Pb)- <0.0050%, Nitrate (NO3)- <0.0030%, Phosphate (PO4)- <0.0005%, Sulfate (SO4) - <0.0200%
Chemical Name or Material Molybdenum Trioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.