missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum rod, 6.4mm (0.25in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 11376727
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
7439-98-7
Mo
95.95
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
23932
CHEBI:28685
molybdenum
[Mo]
This item is not returnable. View return policy
7439-98-7
Mo
95.95
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
23932
CHEBI:28685
molybdenum
[Mo]

Approx. 15.2cm/50g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance Dark gray rod
Assay Percent Range 99.95% (Metals basis)
Quantity 50 g
Odor Odorless
Form Rod
Chemical Name or Material Molybdenum

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'