missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum, plasma standard solution, Specpure™ Mo 10,000μg/mL

Product Code. 11378657
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378657 50 mL 50mL
11368657 100 mL 100mL
11425070 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378657 Supplier Thermo Scientific Alfa Aesar Supplier No. 035766.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00003465

Specifications

Physical Form Liquid
Concentration Mo in 5% HNO3/tr. HF
UN Number UN3264
Quantity 50 mL
Grade Specpure
Chemical Name or Material Molybdenum plasma standard solution, Mo 10000μg/mL

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.