missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(IV) sulfide, 98.5%

Product Code. p-4509981 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10032670

Brand: Thermo Scientific Acros 215781000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1317-33-5 | MoS2 | 160.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-33-5
Molecular Formula MoS2
Molecular Weight (g/mol) 160.07
InChI Key CWQXQMHSOZUFJS-UHFFFAOYSA-N
Synonym molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
PubChem CID 14823
ChEBI CHEBI:30704
IUPAC Name bis(sulfanylidene)molybdenum
SMILES S=[Mo]=S

Specifications

Melting Point 2375.0°C
Color Black or Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 98% min. (MoS2)
Linear Formula MoS2
Merck Index 15,6322
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in aqua regia and sulfuric acid
Formula Weight 160.07
Percent Purity 98.5%
Packaging Glass bottle
Chemical Name or Material Molybdenum(IV) sulfide
Show More Show Less
compliance-icons
Product Identifier
  • Molybdenum(IV) sulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.