missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(III) chloride, 99.5% (metals basis)

Product Code. 11321569
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321569 2 g 2g
11331569 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11321569 Supplier Thermo Scientific Alfa Aesar Supplier No. 014034.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13478-18-7 | Cl3Mo | 202.3 g/mol

Molybdenum(III) chloride reacts with tetrahydrothiophene to form complexes, which catalyze the polymerization and cyclotrimerization of alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-18-7
Molecular Formula Cl3Mo
Molecular Weight (g/mol) 202.3
MDL Number MFCD00014214
InChI Key ZSSVQAGPXAAOPV-UHFFFAOYSA-K
Synonym molybdenum trichloride, molybdenum iii chloride, molybdenum chloride mocl3, mocl3, trichloridomolybdenum, acmc-1bwlx, molybdenum 3+ chloride
PubChem CID 83515
ChEBI CHEBI:30626
IUPAC Name trichloromolybdenum
SMILES Cl[Mo](Cl)Cl

Specifications

Melting Point (decomposition)
Color Red
Physical Form Powder
Quantity 2 g
UN Number UN3260
Solubility Information Soluble in nitric acid and sulfuric acid. Insoluble in water and dilute hydrochloric acid. Slightly soluble in ethanol and diethyl ether.
Formula Weight 202.32
Percent Purity 99.5%
Odor Acidic
Sensitivity Air and moisture sensitive
Assay (metals basis)
Chemical Name or Material Molybdenum(III) chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.