missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(II) acetate dimer

Product Code. 11343437
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343437 1 g 1g
11353437 5 g 5g
11333437 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343437 Supplier Thermo Scientific Alfa Aesar Supplier No. 018239.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14221-06-8 | C8H12Mo2O8 | 428.076 g/mol

Molybdenum(II) acetate dimer is generally used as an intermediate compound in a process to form other quadruply bonded molybdenum compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14221-06-8
Molecular Formula C8H12Mo2O8
Molecular Weight (g/mol) 428.076
MDL Number MFCD00013395
InChI Key DOOLFANBWPPEGQ-UHFFFAOYSA-J
Synonym molybdenum ii acetate, tetrakis mu-acetato-o:o' dimolybdenum, molybdenum 2+ tetraacetate, dimolybdenum tetraacetate, tetra aceto dimolybdenum, molybdenum ii acetate dimer,, molybdenum ii acetate dimer 1g, molybdenum ii acetate dimer
PubChem CID 84269
IUPAC Name molybdenum(2+);tetraacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Mo+2].[Mo+2]

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in organic solvents. Insoluble in water.
Formula Weight 428.06
Physical Form Crystalline
Chemical Name or Material Molybdenum(II) acetate dimer

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.