missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Molybdenum gauze, 165 mesh woven from 0.0509mm (0.002 in.) dia. wire, twilled weave

Brand:  Alfa Aesar™ 042404.FL

Code : NEW

Additional Details : CAS Number : 7439-98-7 Weight : 0.01000kg

Quantity 75x75mm

 View more versions of this product

Product Code. 11374548

  • £161.10 / Each


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Specifications

Chemical Identifiers

Molybdenum gauze
7439-98-7
165 mesh woven from 0.0509mm (0.002 in.) dia. wire, twilled weave
MFCD00003465

Specifications

Molybdenum gauze
165 mesh woven from 0.0509mm (0.002 in.) dia. wire, twilled weave
Wire Cloth
165 Mesh
75x75mm
7439-98-7
MFCD00003465
Videos
SDS