missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum foil, 0.1mm (0.004in) thick, 99.95% (metals basis)

Product Code. p-3257049
Click to view available options
Quantity:
100 x 150 mm
150 x 300 mm
300 x 600 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11385729

Brand: Thermo Scientific Alfa Aesar 010043.GJ

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 10.2g/100x100mm | CAS: 7439-98-7 | Mo | 95.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Color Gray
Appearance Dark gray foil
Density 10.22g/cm3 at 20°C
Quantity 100 x 150 mm
Odor Odorless
Assay Percent Range 99.95% (Metals basis)
Form Foil
Formula Weight 95394
Chemical Name or Material Molybdenum

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.