missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum boride, 99% (metals basis)

Product Code. 11347407
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347407 100 g 100g
11367407 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347407 Supplier Thermo Scientific Alfa Aesar Supplier No. 012563.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12006-98-3 | BMo | 106.76 g/mol

Molybdenum borides have been used as hard coating materials for high temperature structural applications, and also in the form of MoB/CoCr cermet coatings. It is also used as a brazing material especially in electric components. Molybdenum boride has potential industrial applications such as abrasive, corrosion resistant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12006-98-3
Molecular Formula BMo
Molecular Weight (g/mol) 106.76
MDL Number MFCD00014219
InChI Key LGLOITKZTDVGOE-UHFFFAOYSA-N
Synonym molybdenum boride, molybdenum boride mob, molybdenumylidyneborane, molybdenum monoboride, molybdenum boride mono , mob, molybdenum boride, powder,-325 mesh trace metals basis
PubChem CID 82790
IUPAC Name boranylidynemolybdenum
SMILES B#[Mo]

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information it is soluble in water,paraffin oil,ether,benzene and other organic solvents.
Formula Weight 106.75
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Molybdenum boride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.