missing translation for 'onlineSavingsMsg'
Learn More

Molsidomine, MedChemExpress

Product Code. 16472246
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472246 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472246 Supplier Medchem Express Supplier No. HYB1069
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Molsidomine is an orally active, long acting vasodilating drug, metabolized in the liver to the active metabolite linsidomine, which is an unstable compound that releases nitric oxide (NO) upon decay as the actual vasodilating compound.

Molsidomine is an orally active, long acting vasodilating drug, metabolized in the liver to the active metabolite linsidomine, which is an unstable compound that releases nitric oxide (NO) upon decay as the actual vasodilating compound.

Specifications

Chemical Name or Material Molsidomine
CAS 25717-80-0
Color White
Purity Grade Notes Research
Molecular Formula C9H14N4O4
Quantity 500 mg
Synonym SIN-10 Morsydomine
Solubility Information DMSO : ≥ 100 mg/mL (412.83 mM) ∣H2O : 25 mg/mL (103.21 mM; Need ultrasonic)
SMILES O=C([N-]C1=C[N+](N2CCOCC2)=NO1)OCC
Molecular Weight (g/mol) 242.23
Formula Weight 242.23
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.