missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, Powder, Catalyst Support, Sodium Y Zeolite, Honeywell Fluka™

Product Code. 15683970 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15683970 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15683970 Supplier Honeywell Fluka Supplier No. 334448100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Powder, Catalyst support, sodium Y zeolite

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly bottle
Quantity 100 g
CAS 63231-69-6
Physical Form Powder, Catalyst support, sodium Y zeolite
MDL Number MFCD00131613
UN Number NONH for all modes of transport
IUPAC Name Molecular sieves 13X
Chemical Name or Material Molecular sieves
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.