missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 4A, 1-2mm (0.04-0.08in) dia. pellets

Product Code. p-7080393
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11359139

Brand: Thermo Scientific Alfa Aesar 087956.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol

Molecular sieves, 4 Å… is used to absorb molecules as large as butane. It is utilized for the separation of natural gas, liquified petroleum gas (LPG) and alkenes. It finds application in the refining and purification of liquid and gases. It is also used for drying and purification of air, inert gases and solvents. It is actively involved to remove methanol, carbon dioxide and ammonia from fluid streams. It plays a vital role to remove heavy metal ions available in water. It is also used as a soap forming agent and as a detergent auxiliary. It has been considered as a general-purpose drying agent in polar and non polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
MDL Number MFCD00131613
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
IUPAC Name (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Color Brown
Sensitivity Hygroscopic
Quantity 1 kg
Physical Form Pellet
Solubility Information Insoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone.
Chemical Name or Material Molecular sieves 4A, 1 to 2mm (0.04 to 0.08 in.) dia. pellets

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.