missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 3A, Powder, TRC

Product Code. 30467878
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
30467878 500 g 500g
1 options
This item is not returnable. View return policy

Product Code. 30467878

Brand: Toronto Research Chemicals TRCM489003500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Molecular Sieves, 3A, Powder
CAS 308080-99-1
Molecular Formula AlNaO6Si2
Quantity 500 g
Synonym Zeolites, 3A, 3A Zeolites, 3ASDG, Baylith SG 232, HTMS 3A, LEK 2425, MS 3A, Mizuka Sieves 3A, Molecular Sieve 3A, Molsiv 3A, Purmol 3A, Purmol 3STH, S 136, Siliporite 3A, Siliporite NK 30, Siliporite NK 30AP, Sylobead 562CS, Tricat 3A, UOP-AS 5078, Z 3-02, Zeolum A 3
InChI Formula InChI=1S/Al.Na.2O3Si/c;;2*1-4(2)3/q+3;+1;2*-2
SMILES O=[Si]([O-])[O-].O=[Si]([O-])[O-].[Al+3].[Na+]
Molecular Weight (g/mol) 202.14
Formula Weight 201.89
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.