missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves 3A, Pellets, Dia.: 3.2mm, Honeywell Fluka™

Product Code. p-7120591 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Drum with inliner
Poly Bottle
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 15613960

Brand: Honeywell-Fluka 3342781KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pellets, 3.2 mm

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Specifications

Color Beige or Tan
Density 2 g/cm3
Packaging Poly Bottle
Particle Size Dia.: 3.2 mm
Quantity 1 kg
Physical Form Pellets
Melting Point >400°C
Linear Formula KnNa12-n[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
IUPAC Name Molecular sieves 3A
Chemical Name or Material Molecular sieves, UOP Type 3Å
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.