missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 3A, 8 to 12 mesh

Product Code. 10082590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
5 kg
25 kg
Unit Size:
25kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10082590 5 kg 5kg
10082600 500 g 500g
15204674 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10082590 Supplier Thermo Scientific Acros Supplier No. 197250050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Specifications

Color Beige to Brown
Bulk Density 600g/L min.
Packaging Plastic drum
Residual Water 1.5% max.
Water Adsorption 20% min. (60% rel. humid.)
Quantity 5 kg
Physical Form Beads
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Molecular sieves 3A, 8 to 12 mesh

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.