missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 3 Å, 4 to 8mesh, Solstice

Product Code. 15661840
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15661840 1 kg 1kg
15671840 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15661840 Supplier Honeywell-Fluka Supplier No. 2085741KG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
This item is not returnable. View return policy

Beads, 4-8 mesh

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Specifications

Mesh Size 4 to 8mesh
Quantity 1 kg
Linear Formula KnNa12-n[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
Chemical Name or Material Molecular Sieves, 3 Å
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.