missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 13X, powder, Thermo Scientific Chemicals

Catalog Number 11424203
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
63231-69-6
MFCD00131613
Molecular sieves 13X
This item is not returnable. View return policy
63231-69-6
MFCD00131613
Molecular sieves 13X

They are used as catalyze carrier, co adsorption of CO2 and H2O and H2O, H2O and H2S, as desiccant for medical and air compressor systems. They are normally used in gas phase and liquid phase applications. It finds its a major applications in commercial gas drying, air plant feed purification (simultaneous H2O and CO2 removal) and liquid hydrocarbon/natural gas sweetening (H2S and mercaptan removal).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Sensitivity Hygroscopic
Quantity 250 g
Physical Form Powder
Solubility Information Insoluble in water.
Chemical Name or Material Molecular sieves 13X

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'