missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 13X, Pellets, Dia.: 1.6mm, Solstice

Product Code. 15633970 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Drum with inliner
Poly Bottle
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15633970 5 kg Drum with inliner 5kg
15623970 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15633970 Supplier Honeywell-Fluka Supplier No. 3343405KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Pellets, 1.6 mm diameter

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Packaging Drum with inliner
Particle Size Dia.: 1.6mm
Quantity 5 kg
Physical Form Pellets
Linear Formula Na86[AlO2)86(SiO2)106] · xH2O
UN Number NONH for all modes of transport
Chemical Name or Material Molecular sieves, 13X
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.