missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 13X, 4 to 8 mesh

Product Code. 10471431 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10471431 500 g Glass bottle 500g
10617162 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10471431 Supplier Thermo Scientific Acros Supplier No. 197305000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63231-69-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Color Beige to Brown
Bulk Density 600g/L min.
Packaging Glass bottle
Residual Water 1.5% max.
Quantity 500 g
Physical Form Beads
Chemical Name or Material Molecular sieves 13X, 13X, 4 to 8 mesh

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.