missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Molecular sieves, 13X, 3-4mm (0.12-0.16 in.) dia.. pellets

Brand:  Alfa Aesar™ 033550.A1

Code : NEW

Additional Details : CAS Number : 63231-69-6 Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 11337758

  • £48.60 / 1kg


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Specifications

Chemical Identifiers

Molecular sieves 13X
63231-69-6
3 to 4mm (0.12 to 0.16 in.) dia. pellets
MFCD00131613

Specifications

Molecular sieves 13X
Hygroscopic
3 to 4mm (0.12 to 0.16 in.) dia. pellets
Pellet
1/8 in.
1kg
63231-69-6
MFCD00131613
Videos
SDS