missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieve Type 3A, Technical, 1.6-2.5mm Beads (8-12 Mesh), Fisher Chemical™

Product Code. 10070270 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10070270 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10070270 Supplier Fisher Chemical Supplier No. MS/1010/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

AlNaO6Si2, CAS Number-308080-99-1, 500 g, Beige, URGAHOPLAPQHLN-UHFFFAOYSA-N, aluminum;sodium;dioxido(oxo)silane, 131613, 202.135, Plastic powder jar, Solid, 19758701, [O-][Si](-O)[O-].[O-][Si](-O)[O-].[Na+].[Al+3] | CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol

AlNaO6Si2, CAS Number-308080-99-1, 500 g, Beige, URGAHOPLAPQHLN-UHFFFAOYSA-N, aluminum;sodium;dioxido(oxo)silane, 131613, 202.135, Plastic powder jar, Solid, 19758701, [O-][Si](-O)[O-].[O-][Si](-O)[O-].[Na+].[Al+3]

TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Packaging Plastic powder jar
Quantity 500 g
CAS 308080-99-1
Physical Form Solid
Molecular Formula AlNaO6Si2
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]
IUPAC Name aluminum;sodium;dioxido(oxo)silane
Molecular Weight (g/mol) 202.135
PubChem CID 19758701
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.