missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieve Type 13X (10A), Technical, 1.6-2.5mm Beads (8-12 Mesh), Fisher Chemical

Product Code. 10763801 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10763801 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10763801 Supplier Fisher Chemical Supplier No. MS/1070/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Beige
pH 10
Packaging Plastic powder jar
Quantity 500 g
CAS 63231-69-6
Physical Form Solid
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.