missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieve Deperox/Dehydrate, Honeywell Fluka™

Product Code. 15676730 Shop All Honeywell Fluka Products
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1 kg
missing translation for 'unitSize'
1kg
250g
This item is not returnable. View return policy

Product Code. 15676730

missing translation for 'mfr': Honeywell Fluka 69805250G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Beads, for the deperoxidation and drying of solvents

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Molecular Sieve Deperox/Dehydrate Fluka for the deperoxidation and drying of solvents
Packaging Poly bottle
Quantity 250 g
CAS 10124-43-3, 1318-02-1
UN Number UN3077
compliance-icons compliance-icons
Product Identifier
  • Molecular Sieve Deperox/Dehydrate Fluka
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : cobalt sulfate, Zeolite, cuboidal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.