missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieve Dehydrate with Indicator, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Molecular Sieve Dehydrate Fluka with indicator for drying solvents |
| Packaging | Poly bottle |
| Quantity | 250 g |
| UN Number | UN3077 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction