missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieve Dehydrate with Indicator For Drying Gases, Beads, Honeywell Fluka™

Product Code. 15632930 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15632930

Brand: Honeywell Fluka 28211250G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Beads, with indicator for drying gases

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Molecular Sieve Dehydrate Fluka With Indicator For Drying Gases
Density 2g/cm
Packaging Glass Bottle
Pore Size 4
Quantity 250 g
CAS 1318-02-1, 10124-43-3
UN Number NONH for all modes of transport
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.