missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ X-GlcU, CHA (5-Bromo-4-Chloro-3-Indolyl β-D-Glucuronide, Cyclohexylammonium Salt)

Product Code. 11544167
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11544167

Brand: Invitrogen™ B1691

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Widely used chromogenic substrate for β-glucuronidase (GUS)

Yields a dark blue precipitate at the site of enzymatic activity. X-GlcU is routinely used to detect GUS expression in transformed plant cells and tissues. X-GlcU can also be used to detect E. coli contamination in food and water.

Additional Information About Glycosidase Enzymes
Glycosidase enzymes exhibit very high selectivity for hydrolysis of their preferred sugars. For example, β-galactosidase rapidly hydrolyzes β-D-galactopyranosides but usually does not hydrolyze either the anomeric α-D-galactopyranosides or the isomeric β-D-glucopyranosides. Endogenous glycosidase activity is frequently used to characterize strains of microorganisms and to selectively label organelles of mammalian cells; defects in glycosidase activity are characteristic of several diseases.

In addition, glycosidases are important reporter gene markers. Specifically, lacZ, which encodes β-galactosidase, is extensively used as a reporter gene in animals and yeast, whereas the β-glucuronidase (GUS) gene is a popular reporter gene in plants. Glycosidase substrates are also used in conjunction with glycosidase-conjugated secondary detection reagents in immunohistochemical techniques and enzyme-linked immunosorbent assays (ELISAs).

  • Chromogenic substrate
  • Cell permeant
  • Useful to detect GUS expression in transformed plant cells and tissues

Beta-Glucuronidase (GUS) Assays, Cell Analysis, Cellular Imaging, Enzyme and Protein Activity Assays, Glycosidase, Neuraminidase, and Related Enzyme Activity, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunohistochemistry (IHC), Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-Permeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Colorimetric
For Use With (Application) Molecular Biology
Quantity 100 mg
Substrate Beta-Glucuronidase (GUS) Substrate
Substrate Properties Chemical Substrate
Substrate Type Beta-Glucuronidase (GUS) Substrate
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.