missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Molecular Probes™ TSA™ Kit 27, with HRP-streptavidin and Alexa Fluor™ 350 tyramide

Product Code. 10247342
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10247342 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10247342 Supplier Invitrogen™ Supplier No. T20937

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enzyme-mediated detection method uses the catalytic activity of horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid sequence in situ

  • Includes HRP-streptavidin and blue-fluorescent Alexa Fluor 350 tyramide, which is spectrally similar to AMCA dye

Cell Analysis, Cellular Imaging, Fluorescence In Situ Hybridization, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Immunohistochemistry (IHC), In Situ Hybridization (ISH)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Quantity 1 Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.