missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Molecular Probes™ TSA™ Kit 16, with HRP-Goat Anti-Rabbit IgG and Alexa Fluor™ 647 Tyramide

Product Code. 10493252
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10493252 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10493252 Supplier Invitrogen™ Supplier No. T20926

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

HRP-goat anti-rabbit IgG and far-red-fluorescent Alexa Fluor 647 tyramide

Tyramide signal amplification is enzyme-mediated detection method that uses catalytic activity of horseradish peroxidase (HRP) to generate high-density labeling of target protein or nucleic acid sequence in situ

  • Alexa Fluor 647 tyramide is spectrally similar to Cy5 dye

Cell Analysis, Cellular Imaging, Fluorescence In Situ Hybridization, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Immunohistochemistry (IHC), In Situ Hybridization (ISH)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5° to -30°C) and protect from light.
Quantity 1 Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.