missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Molecular Probes™ TSA™ Kit 12, with HRP-Goat Anti-Rabbit IgG and Alexa Fluor™ 488 Tyramide

Product Code. 10144952
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity unitSize
10144952 1 Kit 1 set
1 options
This item is not returnable. View return policy

Product Code. 10144952

Brand: Invitrogen™ T20922

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

HRP-goat anti-rabbit IgG and green-fluorescent Alexa Fluor 488 tyramide

Tyramide signal amplification is enzyme-mediated detection method that uses catalytic activity of horseradish peroxidase (HRP) to generate high-density labeling of target protein or nucleic acid sequence in situ

  • Alexa Fluor 488 tyramide is spectrally similar to fluorescein

Cell Analysis, Cellular Imaging, Fluorescence In Situ Hybridization, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Immunohistochemistry (IHC), In Situ Hybridization (ISH)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5° to -30°C) and protect from light.
Quantity 1 Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.