Learn More
Invitrogen™ Molecular Probes™ TSA™ Kit 1, with HRP-Goat Anti-Mouse IgG and biotin-XX Tyramide
Description
Includes
HRP-goat anti-mouse IgG and biotin-XX tyramide
- Deposited biotin molecules can be detected with avidin, streptavidin or neutravidin conjugates
Cell Analysis, Cellular Imaging, Fluorescence In Situ Hybridization, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Immunohistochemistry (IHC), In Situ Hybridization (ISH)
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer (-5° to °30°C) and protect from light. |
| Quantity | 1 Kit |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.