Learn More
Invitrogen™ Tali™ Viability Kit - Dead Cell Red
Description
Includes
Tali - Dead Cell Red Reagent
The TaliViability Kit - Dead Cell Red is a ready-to-use solution of propidium iodide (PI). PI is a live cell-impermeant fluorogenic DNA-binding dye that has been extensively used to identify necrotic cells. After staining a cell population using Dead Cell Red, dead cells display red fluorescence upon excitation. TaliViability Kit - Dead Cell Red can be used to identify dead cells in a population of unstained cells, or in a population of GFP-expressing cells. Compatible with eukaryotic cells.
- Impermeant to live cells
- Easily enters and binds to nucleic acids in necrotic cells
Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, Imaging Cytometry
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Product Type | Cell Viability Kit |
| Cell Type | Eukaryotic Cell |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Tali™ Image-Based Cytometer |
| Content And Storage | Store at room temperature. |
| Compatible Cells | Eukaryotic Cells |
| Label or Dye | Propidium Iodide |
| Shipping Condition | Room Temperature |
| Format | Vial |
| Product Line | Tali |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.