missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTO™ 83 Orange Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO

Product Code. 10043262
Change view
Click to view available options
Quantity:
250 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10043262 250 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10043262 Supplier Invitrogen™ Supplier No. S11364

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

For use with fluorescence microscopy and flow cytometry

The cell-permeant SYTO 83 orange fluorescent nucleic acid stain exhibits bright, orange fluorescence upon binding to nucleic acids. Because the staining pattern of the SYTO dyes in live cells may vary between cell types, we offer the SYTO Orange Fluorescent Nucleic Acid Stain Sampler Kit (Cat. No. S-11360) to enable researchers to find the most appropriate orange-fluorescent SYTO stain for their system.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Content And Storage Store in freezer at -5°C to -30°C and protect from light.
Excitation Wavelength Range 543 nm
Dye Type Cell-Permeant
For Use With (Equipment) Fluorescence Microscope
Quantity 250 μL
Volume (Metric) 250 μL
Detection Method Fluorescence
Description SYTO™ 83 Orange Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO
Emission 559 nm
Product Line SYTO
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Nucleic Acid Stain
SubCellular Localization Nucleic Acids
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.