Learn More
Invitrogen™ SYTO™ 62 Red Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO
Description
The cell-permeant SYTO 62 red fluorescent nucleic acid stain exhibits bright, red fluorescence upon binding to nucleic acids. Because the staining pattern of the SYTO dyes in live cells may vary between cell types, we offer the SYTO Red Fluorescent Nucleic Acid Stain Sampler Kit (Cat. No. S-11340) to enable researchers to find the most appropriate red-fluorescent SYTO stain for their system.
Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Red |
| Content And Storage | Store in freezer at -5°C to -30°C and protect from light. |
| Excitation Wavelength Range | 649 nm |
| Dye Type | Cell-Permeant |
| For Use With (Equipment) | Fluorescence Microscope |
| Quantity | 250 μL |
| Volume (Metric) | 250 μL |
| Detection Method | Fluorescence |
| Description | SYTO™ 62 Red Fluorescent Nucleic Acid Stain - 5 mM Solution in DMSO |
| Emission | 680 nm |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.