missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Electrophoretic Mobility-Shift Assay (EMSA) Kit, with SYBR™ Green & SYPRO™ Ruby EMSA stains

Product Code. 10686353
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10686353

Brand: Invitrogen™ E33075

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Detects both nucleic acid and protein in the same gel

This fluorescence-based Electrophoretic Mobility Shift Assay (EMSA) Kit provides a fast, easy and quantitative method. This kit uses two fluorescent dyes for detection - SYBR™ Green EMSA nucleic acid gel stain for RNA or DNA detection and SYPRO™ Ruby EMSA protein gel stain for protein detection. The nucleic acids and proteins are stained sequentially in the gel after electrophoresis, so there is no possibility that labeling will interfere with the protein binding being assayed.

Enzyme and Protein Activity Assays, Proteins, Expression, Isolation and Analysis, RNAi, Epigenetics and Non-Coding RNA Research, Transcription Factor Activity and Detection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Assay EMSA (Electrophoretic Mobility Shift Assay)
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Product Type Electrophoretic Mobility-Shift Assay (EMSA) Kit
Product Line SYBR, SYPRO
Quantity 1 kit
Shipping Condition Room Temperature
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Electrophoretic Mobility-Shift Assay (EMSA) Kit, with SYBR Green & SYPRO Ruby EMSA stains
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P391-Collect spillage.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Trichloroacetic acid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.