missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SBFI, Tetraammonium Salt, cell impermeant
Description
- Useful in combination with other fluorescent indicators used to correlate changes in intracellular Na+ with Ca2+ and Mg2+ concentrations, intracellular pH, and membrane potential
- Although selectivity of SBFI for Na+ is less than that of calcium indicators such as fura-2, it is sufficient for detection of physiological concentrations of Na+ in presence of other monovalent cations
- The spectral response of SBFI upon ion binding permit excitation ratio measurements, and this indicator can be used with same optical filters and equipment used for fura-2
- Label (Ex/Em): SBFI (approximately 340,380/500nm)
- Lyophilized product may be dissolved in distilled water or aqueous buffer for use
- Product is typically loaded into cells by diffusion from patch pipet for correlated fluorescence imaging and electrophysiological recording
- Dissociation constant (Kd) of SBFI for Na+ is 3.8mm in the absence of K+, and 11.3 mm in solutions with combined Na+ and K+ concentration of 135 mm (which approximates physiological ionic strength). SBFI is approximately 18-fold more selective for Na+ than for K+
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Sodium and Potassium Detection
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store at room temperature and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | SBFI Tetraammonium Salt |
| Dye Type | Sodium Indicator |
| Quantity | 1 mg |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?