missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rhodamine 110, bis-(CBZ-L-Isoleucyl-L-Prolyl-L-Arginine Amide), Dihydrochloride (BZiPAR)

Product Code. 11560646
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11560646 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11560646 Supplier Invitrogen™ Supplier No. R6505

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sensitive and selective substrate for assaying proteases

This Molecular Probes™ bisamide derivative of rhodamine 110 (R110) is a sensitive and selective substrate for assaying proteases in solution or inside living cells. These fluorogenic substrates contain an amino acid or peptide covalently linked to each of R110's amino groups. Upon enzymatic cleavage, the nonfluorescent bisamide substrate is converted first to the fluorescent monoamide and then to R110, with a further increase in fluorescence.

  • Substrate can be used to continuously measure enzyme activity in cell extracts and purified enzyme preparations using fluorometer or fluorescence microplate reader
  • Substrate name: (CBZ-Ile-Pro-Arg)2-R110
  • Useful to measure: Trypsin
  • Ex/Em of R110 product: (498/521nm)
  • Cell-Permeant

Properties of the R110 Reaction Product

  • Has spectral properties similar to those of fluorescein, making it compatible with standard fluorescein filter sets
  • Signal to background ratios are good because of large extinction coefficient of R110
  • Unlike fluorescein, fluorescence intensity of rhodamine 110 reaction product is constant from pH 3–9

Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-Permeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Solid
Quantity 5 mg
Substrate Caspase Substrate
Substrate Properties Peptide-Based Substrate
Target Enzyme Trypsin
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Shipping Condition Room Temperature

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.