Learn More
Invitrogen™ RediPlate™ 96 EnzChek™ Tyrosine Phosphatase Assay Kit
Description
The RediPlate 96 EnzChek Tyrosine Phosphatase Assay Kit is a ready-to-use, fluorescence-based microplate assay for detecting tyrosine phosphatases (PTPase) and their corresponding modulators and inhibitors. Each RediPlate 96-well microplate has removable lanes, enabling researchers to perform only as many assays as needed for each experiment or to perform high-throughput analysis.
A 384-well RediPlate EnzChek PTPase Assay Kit is also available.
Drug Discovery and Development, Enzyme and Protein Activity Assays, Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation and Analysis, Target and Lead Identification and Validation
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence Intensity |
| For Use With (Application) | Phosphatase Assay |
| For Use With (Equipment) | Microplate Reader |
| Product Type | Phosphatase Assay |
| Dye Type | DiFMU |
| Emission | 358/452 |
| Product Line | EnzChek |
| Quantity | 1 x 96-well plate |
| Shipping Condition | Room Temperature |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.