missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pro-Q™ Emerald 300 Glycoprotein Gel Stain Kit, with SYPRO™ Ruby Protein Gel Stain

Product Code. 10555013
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
9g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10555013 1 kit 9g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10555013 Supplier Invitrogen™ Supplier No. P21855

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

50-fold more sensitive than standard fuchsin staining

Pro-Q Emerald 300 and Pro-Q Emerald 488 Glycoprotein Stain Kits provide the most advanced technology available for detection of glycoproteins in gels and on blots. Gel staining is rapid and very sensitive. In less than three hours, it is possible to detect as little as 300 pg of glycoprotein per band, depending on the degree of glycosylation.

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Detection Location In-Blot Detection, In-Gel Detection
Detection Method Fluorescence
Label or Dye SYPRO Ruby, Pro-Q Emerald 300
Quantity 1 kit
Shipping Condition Room Temperature
Target Molecule Proteins (Glycoproteins)
Product Line PRO-Q, SYPRO
Product Type Glycoprotein Staining Kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.