missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pro-Q™ Diamond Phosphoprotein Gel Staining Kits

Product Code. 10195032
Change view
Click to view available options
Quantity:
1 L Stain, 40 μL Standard Kit
200 mL Stain, 40 μL Standard Kit
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10195032 1 L Stain, 40 μL Standard Kit 1 set
10184652 200 mL Stain, 40 μL Standard Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10195032 Supplier Invitrogen™ Supplier No. MPP33300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards.

The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards. Pro-Q Diamond Phosphoprotein Gel Stain is a fluorescent stain (excitation/emission maxima ∼555/580 nm) that can detect as little as 1–16 ng of phosphoprotein per band with a signal that is linear over three orders of magnitude. The PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins with molecular weights from 14,000 to 116,250 daltons that serve both as molecular weight markers and as positive and negative controls.

The Pro-Q Diamond Phosphoprotein Gel Staining Kit includes Pro-Q Diamond Phosphoprotein Gel Stain and PeppermintStick Phosphoprotein Molecular Weight Standards. Pro-Q Diamond Phosphoprotein Gel Stain is a fluorescent stain (excitation/emission maxima ∼555/580 nm) that can detect as little as 1–16 ng of phosphoprotein per band with a signal that is linear over three orders of magnitude. PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins with molecular weights from 14,000 to 116,250 daltons that serve both as molecular weight markers and as positive and negative controls.

Features include:
Sensitive—detection level down to 1–16 ng of phosphoprotein per band
Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis
Built-in controls—molecular weight marker allows for approximation of protein size and serves as positive and negative control with unique mixture of phosphorylated and non-phosphorylated proteins
Linear signal—exhibits excellent linearity over three orders of magnitude, ideal for quantitation

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Pro-Q™ Diamond Phosphoprotein Gel Stain, 1 L
•PeppermintStick™ Phosphoprotein Molecular Weight Standard, 40 μL
Description Pro-Q™ Diamond Phosphoprotein Gel Staining Kit, 1 L
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye Pro-Q Diamond
Quantity 1 L Stain, 40 μL Standard Kit
Target Molecule Proteins (Phosphoproteins)
Product Line PRO-Q
Product Type Phosphoprotein Gel Staining Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.