missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pro-Q™ Diamond Phosphoprotein Gel Destaining Solution

Product Code. 11559296
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11559296 5 L Each
10246142 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11559296 Supplier Invitrogen™ Supplier No. P33311

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Pro-Q Diamond Phosphoprotein Gel Destaining Solution is a ready-to-use solution premixed for your convenience.

Pro-Q Diamond Phosphoprotein Gel Destaining Solution is a ready-to-use solution premixed for your convenience. It is formulated for use with Pro-Q Diamond Phosphoprotein Gel Stain. Destaining is important for reducing the gel background signal and the signal from nonspecific staining.

Pro-Q Diamond Phosphoprotein Gel Destaining Solution is a ready-to-use solution premixed for your convenience. It is formulated for use with Pro-Q Diamond Phosphoprotein Gel Stain. Destaining is important for reducing the gel background signal and the signal from nonspecific staining.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Description Diamond Phosphoprotein Gel Destaining Solution
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye Pro-Q Diamond
Quantity 5 L
Shipping Condition Room Temperature
Target Molecule Proteins (Phosphoproteins)
Product Line PRO-Q
Product Type Phosphoprotein Gel Destaining Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.