Learn More
Invitrogen™ Molecular Probes™ Press-to-Seal™ Silicone Isolator
Product Code. 10432142
Change view
Click to view available options
:
1 well, 20mm dia., 1.0mm deep
Product Code.
10432142 1 well, 20mm dia., 1.0mm deep
1 options
Product Code. 10432142 Supplier Invitrogen™ Supplier No. P18175
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For versatility and flexibility, isolators can be easily trimmed to custom fit enclosures
  • Available either precut to specific interior dimensions
  • Isolators are autoclavable and noncytotoxic
  • Can be used to isolate cells in culture dishes or to separate specimens on microscope slides
Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining and Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Sample Preparation, Immunohistochemistry (IHC)
TRUSTED_SUSTAINABILITY
Specifications
Type Press-to-Seal™ Silicone Sheet
Wells 1
Description 1 well, 20mm dia., 1.0mm deep
Diameter (Metric) 20 mm