Learn More
Invitrogen™ PeppermintStick™ Phosphoprotein Molecular Weight Standards
Description
PeppermintStick Phosphoprotein Molecular Weight Standards are a mixture of phosphorylated and nonphosphorylated proteins ranging from 14.4 to 116 kDa. Separation by polyacrylamide gel electrophoresis resolves the standard mixture into two phosphorylated and four nonphosphorylated protein bands.
Applications
- Accurate protein sizing on SDS-polyacrylamide gels and western blots.
- Positive and negative controls for detection of phosphorylated proteins, such as the methods provided by our Pro-Q™ Diamond phosphoprotein gel stain
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Size Range | 14.4 to 116.25 kDa |
| Gel Compatibility | Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels |
| Quantity | 400 μL |
| Product Line | PeppermintStick |
| Stain Type | Fluorescent |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.