missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ovalbumin, Alexa Fluor™ 594 Conjugate
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
The red fluorescent Alexa Fluor 594 ovalbumin conjugate is a protein with a relatively low molecular weight (∽45,000 daltons).
Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 2 mg |
| Conjugate | Alexa Fluor 594 |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label Type | Alexa Fluor Dyes |
| Shipping Condition | Room Temperature |
| Protein Subtype | Albumin Proteins |
| Product Line | Alexa Fluor |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction