missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Molecular Probes™ NBD-X (6-(N-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)amino)Hexanoic Acid)

Product Code. 11589126
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11589126

Brand: Invitrogen™ N316

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Functional analog of dinitrophenyl hapten

The carboxylic acid of NBD-X (6-(N-(7-nitrobenz-2-oxa- 1,3-diazol-4-yl)amino) hexanoic acid) can be activated in an aqueous solution with a water-soluble carbodiimide such as EDAC (E2247) in order to couple this environment sensitive fluorophore to amines, hydrazines or hydroxylamines on biomolecules of interest. NBD fluorescence is quenched upon binding to anti-dinitrophenyl (anti-DNP) antibodies.

Cell Analysis, Cell Structure, Membranes (General) & Lipids

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Quantity 100 mg
Target Cell Membranes, Lipids
Color Green

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.