Learn More
Invitrogen™ Muscimol, BODIPY™ TMR-X Conjugate
Description
Muscimol is a powerful agonist of the GABAA receptor and has been widely used to reversibly inactivate localized groups of neurons. The BODIPY TMR-X muscimol conjugate, can be used, in place of radioactive reagents and methods, to map the distribution of muscimol in the nervous system, as well as to detect the presence of GABAA receptors on cell surfaces.
To make a concentrated stock solution, dissolve in either H2O, buffer or normal saline (∼1 mM) or high quality anhydrous DMSO (∼2 mM). DMSO stock solutions should be stored frozen, desiccated and protected from light. Aqueous stock solution should be used fresh.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 1 mg |
| Shipping Condition | Room Temperature |
| For Use With (Equipment) | Fluorescence Microscope |
| Detection Method | Fluorescence |
| Product Line | BODIPY |
| Molecular Formula | C31H36BF2N5O5 |
| Physical Form | Solid |
| Color | Yellow |
| Product Type | Agonist |
| Excitation/Emission | 543/572 nm |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.