missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-Hexadecanoylaminofluorescein

Product Code. 11504876
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11504876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11504876 Supplier Invitrogen™ Supplier No. H110

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lipophilic

5-hexadecanoyl fluorescein binds to membranes with the fluorophore at the aqueous interface and the alkyl tail protruding into the lipid interior.

Cell Analysis, Cell Structure, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Membranes (General) & Lipids, Plasma Membrane

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mg
Shipping Condition Room Temperature
Sub Cellular Localization Cell Membranes & Lipids
Product Line Molecular Probes
Label Type FITC (Fluorescein)
Physical Form Solid
Product Type 5-Hexadecanoylaminofluorescein

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.