missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Low Density Lipoprotein from Human Plasma, Acetylated (AcLDL)

Product Code. 10023522
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10023522 200 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10023522 Supplier Invitrogen™ Supplier No. L35354

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Acetylated (Ac) apoprotein used to study cell types that express receptors specific for this acetylated version, i.e., endothelial and microglial cells

Human low-density lipoprotein (LDL) is large protein complex (approx. 500,000 Da) that binds to a specific receptor on surface of vertebrate cells and delivers cholesterol via receptor-mediated endocytosis. Experiments to study LDL are typically performed by adding fluorescently labeled LDL to cultured cells and analyzing them by microscopy or flow cytometry. Alternatively the fluorescently labeled LDL can be injected into test animals, and the distribution of the label can be analyzed after the specified time period.

  • Acetylated: Yes
  • Amount: 200μL (1.0mg/mL)
  • DL and AcLDL products from fresh human plasma approximately every two months are available on continuous basis
  • LDL containing unmodified apoprotein is used to study normal cholesterol delivery and internalization: If the lysine residues of LDL apoprotein have been acetylated, the LDL complex no longer binds to the LDL receptor, but rather is taken up by endothelial and microglial cells that possess ;scavenger receptors specific for that modified form
  • Applications for labeled LDL complexes include: Counting cell-surface LDL receptors, and analyzing their motion and clustering following internalization, Quantitating LDL receptor activity in fibroblasts (replacing the radiolabeled LDL assay), Investigating LDL expression and identifying LDL receptor deficiencies in various cell lines

Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2.5 mg⁄ml
Content And Storage Store in refrigerator (2–8°C).
Detection Method Fluorescence
Product Type Supplement
Dye Type Unlabeled
Form Liquid
Quantity 200 μL
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.