missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCS LipidTOX™ Phospholipidosis and Steatosis Detection Kit, for high-content screening, for cellular imaging

Product Code. 10125632
Change view
Click to view available options
Quantity:
1200 Assays
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10125632 1200 Assays 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10125632 Supplier Invitrogen™ Supplier No. H34158

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Phospholipid and neutral lipid stains, nuclear stain Hoechst 33342, propranolol and cyclosporin A (control compounds), DMSO.

To characterize phospolipidosis and steatosis

HCS LipidTOX Phospholipidosis and Steatosis Detection Kit offers a complete set of reagents for performing image-based high-content screening (HCS) assays to characterize phospolipidosis and steatosis, toxic side effects on lipid metabolism that can be triggered by drugs or other compounds. The kit supplies LipidTOX Red phospholipid stain and LipidTOX Green neutral lipid stain, which can be used sequentially for the analysis of phospholipidosis and steatosis, respectively, or can be used in isolation for single-parameter analysis. Both fluorescent probes can be easily detected with fluorescence microscopes and can be quantified with stand-alone image analysis software or the built-in image analysis software of most HCS readers.

Cell Analysis, Cellular Imaging, High-Content Screening (HCS)

Order Info

Shipping Condition: Room temperature. Sufficient reagents are supplied for two 96-well plates or ten 96-well plates.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage When stored at -20°C, desiccated, and protected from light, the kit is stable for at least 6 months.
Detection Method Fluorescence
For Use With (Equipment) High Content Analysis Instrument
Product Type Phospholipidosis and Steatosis Detection Kit
Dye Type Other Label(s) or Dye(s)
Format 96-well plate
Product Line LipidTOX, Molecular Probes
Quantity 1200 Assays
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.