Learn More
Invitrogen™ Lectin PNA From Arachis hypogaea (peanut), Alexa Fluor™ 594 Conjugate
Description
Lectin PNA will agglutinate human erythrocytes, but only after neuraminidase treatment.[ALTREF PN40265]
Cell Analysis, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Cellular Imaging, Endocytosis, Exocytosis & Phagocytosis, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbial Tracking
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 1 mg |
| Source | Peanut |
| Conjugate | Alexa Fluor 594 |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label Type | Alexa Fluor Dyes |
| Shipping Condition | Room Temperature |
| Form | Lyophilized |
| Protein Subtype | Lectins |
| Product Line | Alexa Fluor |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.